Learn More
BD Cytometric Bead Array (CBA) Human IgE; Flex Set, Bead E9
Bead-based immunoassay for measuring human total immunoglobulin E (IgE) in serum and plasma samples
Brand:  BD 558682
Product Code. 15819578
  • 620.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human IgE, Standard (two vials), Human total IgE PE Detection Reagent (one vial), Human total IgE Capture Bead E9 (one vial)
Description
Description
  • Human reactivity was determined by testing samples with IgE Flex Set
  • Theoretical limit of detection is 0.07ng/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgE
Human IgE, standard (2 vials), Human total IgE PE detection reagent (one vial), Human total IgE capture bead E9 (one vial)
4°C
Human IgE flex set (bead E9)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research use only