Learn More
BD Cytometric Bead Array (CBA) Human IgG2; Flex Set, Bead C5
Bead-based immunoassay capable of measuring human immunoglobulin G subclass 2 (IgG2) in serum and plasma samples
Brand: BD 558676
Alert:
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes:
- Standard
- PE Detection Reagent
- Capture Bead C5
Description
The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and when reconstituted in 1.0mL Assay Diluent has an IgG2 protein concentration of 3400ng/mL.
- Human reactivity was determined by testing samples with CBA Human IgG2 Flex Set
- Must be used in conjunction with CBA Human Immunoglobulin Master Buffer Kit, flow cytometer and FCAP Array Software
- Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard
Specifications
IgG2 | |
standard, PE detection reagent, capture bead C5 | |
4°C | |
Human IgG2 flex set (bead C5) |
Human | |
100 Tests | |
Flow Cytometry |