Learn More
BD Cytometric Bead Array (CBA) Human MIG; Flex Set, Bead E8
Bead-based immunoassay capable of measuring human monokine-induced by interferon-γ (MIG), also known as CXCL9, in serum, plasma, and cell culture supernatant samples
Brand:  BD 558286
Product Code. 15889278
  • 645.00 € / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human MIG; Standard; Human MIG; PE Detection Reagent; Human MIG; Capture Bead E8
Description
Description
Human reactivity was determined by testing samples with the BD CBA Human MIG Flex Set.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MIG
Capture bead, detection reagent and standard
Serum, Cell Culture Supernatant
Human MIG flex set (bead E8)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Human MIG Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT