Learn More
BD Cytometric Bead Array (CBA) Human TNF; Flex Set, Bead C4
Human and non-human primate reactivity was determined by testing samples with the BD CBA Human TNF Flex Set
Brand:  BD 560112
Product Code. 15859938
  • 655.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human TNF; Standard; Human TNF; PE Detection Reagent; Human TNF; Capture Bead C4
Description
Description
  • Capable of measuring human tumor necrosis factor (TNF), formerly known as tumor necrosis factor-α, in serum, plasma, and cell culture supernatant samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNF
Human TNF, standard, Human TNF, PE detection reagent, Human TNF, capture bead C4
4°C
Human TNF flex set (bead C4)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only