Learn More
BD Cytometric Bead Array (CBA) Human TNFRI ; Flex Set, Bead C4
Bead-based immunoassay for measuring soluble human type 1 tumor necrosis factor receptor (TNFRI), in serum, plasma and cell culture supernatant sample
Brand:  BD 560156
Product Code. 15899968
  • 1025.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human TNFRI Standard; Human TNFRI PE Detection Reagent; Human TNFRI Capture Bead C4
Description
Description
Human reactivity was determined by testing samples with the BD™ CBA Human TNFRI Flex Set.
  • Reactivity: QC Tested: Human
  • Size: 100 tests
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNFRI
Human TNFRI standard, Human TNFRI PE detection reagent, Human TNFRI capture bead C4
4°C
Human soluble TNFRI flex set (bead C4)
Human
100 Tests
Flow Cytometry