Learn More
BD Cytometric Bead Array (CBA) Human TNFRII; Flex Set, Bead C5
Human reactivity was determined by testing samples with the BD CBA Human TNFRII Flex Set
Brand:  BD 560155
Product Code. 15889968
  • 905.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human TNFRII; Standard; Human TNFRII; PE Detection Reagent; Human TNFRII; Capture Bead C5
Description
Description
  • Capable of measuring soluble human type 2 tumor necrosis factor receptor (TNFRII), also known as CD120b, in serum, plasma, and cell culture supernatant samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNFRII
Human TNFRII, standard, Human TNFRII, PE detection reagent, Human TNFRII, capture bead C5
4°C
Human soluble TNFRII flex set (bead C5)
Human
100 Tests
Flow Cytometry