missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human VEGF; Flex Set, Bead B8
For detecting human vascular endothelial growth factor in serum, plasma, and cell culture samples
Brand: BD 558336
Includes: Human VEGF; Standard; Human VEGF; PE Detection Reagent; Human VEGF; Capture Bead B8
Description
The BD CBA Human VEGF Flex Set is a bead-based immunoassay capable of measuring human vascular endothelial growth factor (VEGF) in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human VEGF Flex Set.
- Reactivity: Human
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
VEGF | |
Human VEGF, standard, Human VEGF, PE detection reagent, Human VEGF, capture bead B8 | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Human | |
100 Tests | |
4°C | |
Human VEGF flex set (bead B8) |
Research Use Only