missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse MIG; Flex Set, Bead D9
For detecting mouse monokine induced by gamma interferon in serum and cell culture samples
Brand: BD 558341
Includes: Mouse MIG; Standard; Mouse MIG; PE Detection Reagent; Mouse MIG; Capture Bead D9
Description
The BD CBA Mouse MIG Flex Set is a bead-based immunoassay capable of measuring mouse monokine induced by gamma interferon (MIG), also known as CXCL9, in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse MIG Flex Set.
- Reactivity: Mouse
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
MIG | |
Mouse MIG, standard, Mouse MIG, PE detection reagent, Mouse MIG, capture bead D9 | |
4°C | |
Mouse MIG flex set (bead D9) |
Mouse | |
100 Tests | |
Flow Cytometry |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Mouse MIG Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT
Research Use Only