Learn More
BD Cytometric Bead Array (CBA) Mouse RANTES; Flex Set, Bead D8
A bead-based immunoassay capable of measuring mouse RANTES, also known as CCL5, in serum and cell culture supernatant samples
Brand:  BD 558345
Product Code. 15869318
  • 680.00 € / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Includes: Mouse RANTES; Standard; Mouse RANTES; PE Detection Reagent; Mouse RANTES; Capture Bead D8
Description
Description
Mouse reactivity is determined by testing samples with the BD CBA Mouse RANTES Flex Set.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RANTES
Mouse RANTES, standard, Mouse RANTES, PE detection reagent, Mouse RANTES, capture bead D8
Serum, Cell Culture Supernatant
Flow Cytometry
Mouse
100 Tests
4°C
Mouse RANTES flex set (bead D8)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only