Learn More
BD Difco™ Antisera for Serological Confirmation of E. coli O157:H7
E. ColiO Antiserum O157 and E. Coli H Antiserum H7 are used for identifying Escherichia coli O157:H7
134.00 € - 242.00 €
Specifications
Certifications/Compliance | CE Marked |
---|---|
Content And Storage | Refrigerate, store at 2°C to 8°C |
Product Type | Antiserum |
For Use With (Application) | For tube test for serological confirmation of E. coli O157:H7 |
Description
E. coli O157:H7 is an enteric pathogen that requires only a low inoculum to cause disease. Transmission is usually via food items. Serotyping of the entero-hemorrhagic E. coli is useful in the epidemiological documentation of the spread of a particular strain in a foodborne outbreak. At the time of use, test both positive and negative control cultures to check performance of the antiserum.
- Product type: antigens and antisera
- Groupings: D through K Products
- Type: antisera
- Sub type: E.coli - Lyophilized, re-hydrate to 3mL
- Shelf Life: 1095 days
- Label has Natural Rubber Latex statement
Certifications
CE marked
Specifications
CE Marked | |
Refrigerate, store at 2°C to 8°C | |
Antiserum | |
For tube test for serological confirmation of E. coli O157:H7 |
Safety and Handling
missing translation for 'shelfLife' : 1095 Days
- BD Difco E. Coli H Antiserum H7, 3 mL
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.