missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Bordatella pertussis Serotyping Antisera

Rabbit serum for slide agglutination

Brand:  BD 223091


Product Code. 11792764

  • 198.00 € / 1mL
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Contains Mercury. Dispose of properly.

This item is not returnable. View return policy

Description

Description

  • Available in 1.0mL packaged vials

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Marked
Refrigerate, store at 2°C to 8°C
Bordatella pertussis
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vial B. Pertussis Antiserum 1Ml

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.