Learn More
BD Difco™ Bordetella spp. Antigen and Antisera
Quick reactive Gold standard antisera-typing panels for Bordetella Pertusis antigen and Bordetella Parapertusis antisera
267.00 € - 331.00 €
Specifications
Certifications/Compliance | CE Marked |
---|
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
11712774
|
BD
223101 |
1 mL | Bordetella parapertussis Antiserum |
331.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11702844
|
BD
225851 |
5 mL | Antisera |
267.00 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Product type: Antigens and Antisera
- Groupings: A through B products
- Sub type: Quality control antigens
- Type: Antigens
- Quantity: 5mL
- Shelf Life: 730 days
- Positive Control for Bordetella pertussis antiserum
- Subtype: Bordetella Antisera
- Type: Antisera
- Lyophilized, rehydrate to 1mL
- Shelf Life: 965 days
- Rabbit serum for slide agglutination
Certifications
CE marked
Specifications
CE Marked |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P330-Rinse mouth.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to an appropriate treatment and disposal facility in accordancewith applicable laws and regulations, a.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.