missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium
For cultivating recombinant strains of Escherichia coli. BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium is part of a wide selection of dehydrated culture media.
Brand: BD 244020
This item is not returnable.
View return policy
Description
- Formulation: Pancreatic digest of Casein 16g/L, Yeast extract 10g/L, Yeast extract 5g/L
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
<30°C | |
2XYT | |
Pancreatic digest of Casein (Tryptone) 16g/L, Yeast extract 10g/L, NaCl 5g/L | |
Bottle |
Dehydrated | |
Microbiology Media | |
Cultivation of recombinant strains of Escherichia coli | |
500 g |