missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
Description
Certifications
ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, BAM, CCAM, ISO, USDA |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Peptone Water |
| Product Type | Peptone Water |
| Formulation | Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L |
| For Use With (Application) | Isolation of Salmonella species |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction