missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Granulated Agar
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
Brand: BD 214530
This item is not returnable.
View return policy
Description
Applications:
- Solidifying agent for culture media
- Granulated
Specifications
ISO/DIS, 11014 | |
Agar | |
General bacteriological purposes | |
500 g |
1.5% | |
Dehydrated Granule | |
Container |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction