missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani)

Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Brand:  BD 244510

 View more versions of this product

Product Code. 11748902

  • 463.00 € / 2kg
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCAM
Dehydrated
Agar
Propagation and maintenance of Escherichia coli for molecular biology
2000 g
<30°C
LB Agar, Miller
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L
Pail
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.