missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ YPD Agar

Used for maintaining and propagating yeasts. BD™ Difco™ YPD Agar is a dehydrated powder for preparing culture media in molecular microbiology procedures.

251.00 €

Specifications

Color Beige
Content And Storage <30°C
Form Dehydrated Powder
Media Type YPD Agar
Product Type Agar
View More Specs

Products 1
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
11728882
View Documents Promotion Details
BD
242720
500 g Bottle
251.00 €
500g
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
 
Description

Description

General methods in yeast genetics specify using yeast extract-peptone-dextrose (YPD) medium for cultivating Saccharomyces cerevisiae and other yeasts. Yeasts grow well on a minimal medium containing only dextrose and salts. The addition of protein and yeast cell extract hydrolysates allows faster growth so that during exponential or log-phase growth, the cells divide every 90 minutes.

  • Peptone acts as a source of carbon, nitrogen, vitamins and minerals
  • Yeast extract supplies B-complex vitamins, which stimulate bacterial growth
  • Dextrose is the carbohydrate source
  • Agar acts as the solidifying agent
  • pH (6.5% solution, 25°C): 6.5 ± 0.2

Order Info

Prices are for units of one each unless otherwise specified.

Specifications

Specifications

Beige
Dehydrated Powder
Agar
For maintaining and propagating Yeasts in molecular microbiology procedures
<30°C
YPD Agar
Yeast extract 10g/L, Peptone 20g/L, Dextrose 20g/L, Agar 15g/L
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1825 Days

SDS
Documents

Documents

Product Certifications