Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
116.00 € - 720.00 €
Specifications
Certifications/Compliance | For in vitro diagnostic use |
---|---|
Product Type | Antiserum |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Description | Price | Quantity & Availability | ||||
11762734
|
BD
222521 |
1 mL | N. Meningitidis Antiserum Group Z prime (Z' ) | N/A | |||||
11748333
|
BD
222301 |
1 mL | Neisseria meningitidis Antiserum Group C |
670.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11738333
|
BD
222291 |
1 mL | Neisseria meningitidis Antiserum Group B |
720.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11758333
|
BD
222311 |
1 mL | Neisseria meningitidis Antiserum Group D |
209.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11728333
|
BD
222281 |
1 mL | Neisseria meningitidis Antiserum Group A |
161.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11768333
|
BD
222321 |
1 mL | Neisseria meningitidis Antiserum Poly, Groups A, B, C, D | N/A | |||||
11782734
|
BD
222531 |
1 mL | Neisseria meningitidis Antiserum Group W135 |
116.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11702904
|
BD
228801 |
1 mL | N. Meningitidis Antiserum Group X |
179.00 €
5mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11712904
|
BD
228811 |
1 mL | Neisseria meningitidis Antiserum Group Y |
140.00 €
1mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11772904
|
BD
229101 |
3 mL | Neisseria meningitidis Antiserum Poly 2, Groups X, Y, Z |
705.00 €
5mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11742904
|
BD
228911 |
3 mL | Neisseria meningitidis Antiserum Group Z |
222.00 €
5mL |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
For in vitro diagnostic use | |
Antiserum |
Safety and Handling
- BD Difco Neisseria Meningitidis Antiserum Group Z', Bottle, 1 ml
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
For in vitro diagnostic use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.