missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella H Antiserum 1 Complex

Qualitative tube agglutination tests for serological differentiation of Salmonella

Brand:  BD 222721


Product Code. 11782744

Alert:

The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.

This item is not returnable. View return policy

Description

Description

  • Lyophilized and polyclonal rabbit antisera
  • Absorbed or unabsorbed specifically for either phase 1 or phase 2 antigens
  • Perform between 150-1500 tests
  • Identification of Salmonella by flagellar (H) antigens
  • Sodium azide (0.2%) used as preservative

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antiserum
3 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vial Sal H Antiserum 1 Complex

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers