missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Vibrio cholerae Antisera
Quick reactive Gold standard Vibrio cholerae Antisera
122.00 € - 189.00 €
Specifications
Certifications/Compliance | CE Marked |
---|---|
Content And Storage | Refrigerate, store at 2°C to 8°C |
Product Type | Microbiology Media |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11792784
|
BD
224301 |
Microbiology Media |
189.00 €
3mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
11722794
|
BD
224311 |
Microbiology Media |
175.44 €
3mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
11742794
|
BD
224321 |
Microbiology Media |
122.00 €
3mL |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
Description
- Product type: Antigens and Antisera
- Groupings: U through Z Products
- Sub type: Vibrio cholerae
- Type: Antisera
- Lyophilized, rehydrate to 3mL
- Sufficient reagent for 20 slide tests
- Available with 3 different antiserum types and O Antibodies
- Shelf Life: 1095 days
Rabbit serum for slide agglutination test
Certifications
CE marked
Specifications
CE Marked | |
Refrigerate, store at 2°C to 8°C | |
Microbiology Media |
Safety and Handling
missing translation for 'shelfLife' : 1095 Days
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BD Difco Vibrio Cholerae Antiserum Inaba, 3 mL
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title