missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD DrySlide™ Reagent Slides
missing translation for 'orderingAttributeHoverText'
Quantity:
15 Pack
25 Pack
75 Pack
Type:
Indole Reagent
Nitrocefin Reagent
Oxidase Reagent
PYR Kit
missing translation for 'unitSize'
Pack of 15
Pack of 25
Pack of 75
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Informations sur la commande
Slides are individually wrapped and supplied in convenient dispenser packs
Spécification
Spécification
| Type | Indole Reagent |
| Quantity | 75 Pack |
| Certifications/Compliance | CE |
| For Use With (Application) | Used for determining the indole reaction of bacteria |
| Storage Requirements | 2° to 8°C |
Sécurité et traitement
Identifiant produit
- BD BBL DrySlide Indole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 4-Dimethylaminobenzaldehyde
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu