missing translation for 'onlineSavingsMsg'
Learn More

BD GolgiPlug™ Protein Transport Inhibitor (containing Brefeldin A)

Enhance the detectability of cytokine-producing cells with immunofluorescent staining and flow cytometric analysis

Brand:  BD 555029

95.33 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15847968

  • 247.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Block the intracellular protein transport processes when added to in vitro- or in vivo-stimulated lymphoid cells

  • Produce an accumulation of cytokines and/or proteins in the Golgi complex
  • Enhance the detectability of cytokine-producing cells with immunofluorescent staining and flow cytometric analysis
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Recommended for use at 1:1000 in cell culture.
7.2 to 7.6
BD GolgiPlug Protein Transport Inhibitor with Brefeldin A
Liquid
Enhance the detectability of cytokine-producing cells with immunofluorescent staining and flow cytometric analysis
Store undiluted at 4°C
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD GolgiPlug™ Protein Transport Inhibitor (containing Brefeldin A) > BD GolgiPlug Protein Transport Inhibitor with Brefeldin A

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.