missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Mouse Anti-Human CD38

CD38 is expressed by other cells including monocytes, macrophages, dendritic cells, NK cells, myeloid and erythroid precursors and some epithelial cells. BD Horizon™ BV480 Mouse Anti-Human CD38 antigen acts as an ectoenzyme that catalyzes the synthesis and hydrolysis of a Ca++ mobilizing agent, cyclic ADP-ribose.

Brand:  BD 566186

 View more versions of this product

Product Code. 15804829

  • 344.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BV480 Mouse Anti-Human CD38
Flow Cytometry
BV480
Store undiluted at 4°C
25 Tests
QC Testing: Human
5 μL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers