Learn More
BD Human Cell Surface Marker Screening Panel
For screening cell lines, primary cells or tissue, and is compatible with flow cytometry and bioimaging technology platforms
Brand:  BD 560747
Product Code. 15840459
  • 6910.00 € / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Includes: Three 96 well plates, each well containing 2.75μg of antibody, enough for five tests (0.5μg/test) along with AlexaFluor™ 647 conjugated goat anti-mouse Ig and goat anti-rat Ig secondary antibodies
Description
Description
Bioimaging, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only