missing translation for 'onlineSavingsMsg'
Learn More

BD Human intracellular CytoKine-2 Positive Control Cells, HiCK-2

Contains fixed, non-permeabilized human lymphoid cells

Brand:  BD 555062

Product Code. 15837988

  • 1485.00 € / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket

Includes: 5x106 activated human PBMCs (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL (10% dimethylsulfoxide and 90% fetal bovine serum)
This item is not returnable. View return policy

Description

Description

  • Includes cells that express detectable levels of intracellular IL-3, IL-4, IL-10,IL-13 and GM-CSF
  • Prepared by stimulating human PBMCs in the presence of a protein transport inhibitor
  • Cells harvested were fixed and then stored in 1mL of 10% dimethylsulfoxide and 90% fetal bovine serum at -80°
  • Concentration: 5x106 cells/mL

Intracellular staining, Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light Brown
Frozen
IL-3, IL-4, IL-10,IL-13 and GM-CSF
≈5 x 106 Activated Human PBMCs (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL
-80°C
5 x 106 cells/mL
Human
Intracellular staining
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.