Learn More
BD Human T Cell Activation Kit
Reliably determines the level of key phosphorylated signaling proteins involved in T-cell activation
Brand:  BD 560750
Product Code. 15870459
  • 2945.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Alexa Fluor™ 647 Conjugates: p38 MAPK (pT180/Y182), ERK (pT202/204), Stat1 (pY701), Stat3 (pY705), Stat5 (pY694), and Stat6 (pY641) Human T Cell Cocktail, Compensation Reagents, Control Cells and buffers
Description
Description
  • Ready to use
  • Supplied with pre-titrated fluorochrome conjugated antibodies, optimized buffers, instrument setup beads, lyophilized human control cells, validated protocols
  • Includes access to Cytobank software for storing, sharing and analysis of data
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for diagnostic or therapeutic procedures.