Learn More
BD Human Th1/Th17 Phenotyping Kit
Simultaneously view Th1 and Th17 cells in mouse model systems
Brand:  BD 560752
Product Code. 15880459
  • 1010.00 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Includes: Human Th1/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, IFN-γ FITC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to successfully analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for diagnostic or therapeutic procedures.