Learn More
BD Human Th1/Th2/Th17 Phenotyping Kit
Simultaneously view Th1, Th2 and Th17 cells in mouse model systems
Brand:  BD 560751
967.28 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15196713
  • 970.00 € / Each
Explore more special offers
Includes: Human Th1/Th2/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, IFN-γ FITC, IL-4 APC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samplesor to monitor T helper cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • BD Pharmingen Human Th1/Th2/Th17 Phenotyping Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P370+P378-In case of fire: Use water spray, fog, CO2, dry chemical, oralcohol-resistant foam to extinguish.
  • P403-Store in a well-ventilated place.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Monensin, monosodiumsalt, formaldehyde
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for diagnostic or therapeutic procedures.