Learn More
BD Human Th17/Treg Phenotyping Kit
Simultaneously view TH17 and regulatory T cells in human model systems
Brand:  BD 560762
986.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15870469
  • 990.00 € / Each
Explore more special offers
Includes: Human Th17/Treg Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, FoxP3 Alexa Fluor™ 647), Human FoxP3 Buffer A (10X), Human FoxP3 Buffer B (50X), BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor CD4+ T cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Human Th17/Treg Phenotyping Kit > 1 Kit (50 tests)
For research use only. Not for diagnostic or therapeutic procedures.