Learn More
BD IMag™ Buffer
For the optimal labeling of leukocytes using BD IMag particles and for optimal magnetic cell separation using the BD IMagnet™ Cell Separation Magnet
Brand:  BD 552362
Product Code. 15951250
  • 261.00 € / Each
Explore more special offers
Description
Description
A 10X solution containing bovine serum albumin, EDTA, and sodium azide in phosphate buffered saline (PBS).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
Magnetic Cell Separation
4°C