missing translation for 'onlineSavingsMsg'
Learn More

BD Integrins Sampler Kit

For routine testing of western blot analysis

Brand:  BD 611435

Product Code. 15845929

  • 2020.00 € / 1 set
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Fibronectin, Integrin a2/VLA-2a, Integrin a3/VLA-3a, Integrin a5, Integrin aL/LFA-1a, Integrin aV / CD51, Integrin b1, Integrin b3, Integrin b4
This item is not returnable. View return policy

Description

Description

  • Format: Purified
  • Storage buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only