missing translation for 'onlineSavingsMsg'
Learn More

BD Macro-Vue™ RPR (Rapid Plasma Reagin) 18mm Circle Card Tests: Antigen Suspension

Nontreponemal testing procedure for the serologic detection of syphilis

Brand:  BD 270333


 View more versions of this product

Product Code. 17352451

  • 324.00 € / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.

This item is not returnable. View return policy

Description

Description

RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.

  • Recommended when venous blood collection employed and a large volume of serum available, such as in public health and clinical laboratories
  • Various RPR Card Test kits are available with all the necessary supplies to perform the procedure
  • A wide array of kit sizes are available
  • Antigen and testing components may be purchased separately
  • Test cards are available in 10, 15 and 30 spot cards
  • BD Dispenstirs™ are included with tests to serve as pipetting and spreading devices
  • When specimen contains antibody, flocculation occurs with coagglutination of carbon particles of RPR Card antigen, which appear as black clumps against white background of card
  • Nonreactive specimens appear to have an even light-gray color
  • In special situations when nontreponemal test results are needed rapidly and specimen is collected as EDTA plasma, RPR 18mm Circle Card Test can be used if test is performed within 24 hours

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reagin (Syphilis)
Cardiolipin 0.003%, Lecithin 0.020 to 0.022%, Cholesterol 0.09%, EDTA 0.0125M, Na2HPO4 0.01M, KH2PO4 0.01M, Sodium Azide 0.2%, Charcoal 0.02%, Choline Chloride 10% w/v, Deionized/Distilled Water
3mL ampule 3/sp
Antigen Suspension
Serologic detection of Syphilis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD BBL RPR Card Antigen Suspension, 3 ml

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.