Learn More
BD Microtainer™ MAP Microtube for Automated Process with K2EDTA and BD Microgard™ Closure
Designed for the collection, transport and automated processing of skin puncture samples from infants, children, geriatric, oncology and critical care patients
Brand:  BD 363706
Product Code. 12997646
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The BD Microtainer MAP tube is designed to improve turnaround time, safety and accuracy of capillary blood collection and testing in patients with limited blood volume such as infants, children, oncology patients and the elderly.
  • Streamlines operations by allowing for automated processing of your micro-collection hematology samples
  • Enhances patient safety by accommodating full-size patient identification labels at the time of collection
  • Improves workflow with one-piece design, no parts to assemble, no contaminated parts to discard
  • Latex-free, disposable and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
1.0 mg K2 EDTA
250 to 500 μL
Lavender
MAP MicroTube for Automated Process
Non-sterile
Yes
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.