Learn More
BD Mouse Th1/Th2/Th17 Phenotyping Kit
Simultaneously view Th17, Th2 and Th17 cells in mouse model systems
Brand:  BD 560758
Product Code. 15840469
  • 1245.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Mouse Th1/Th2/Th17 Phenotyping Cocktails (CD4 PerCP-Cy™5.5, IL-17A PE, IFN- γFITC, IL-4 APC), BD Cytofix™ Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation
  • Ensures indentification and characterization of the nature of the T helper cell types
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for diagnostic or therapeutic procedures.