missing translation for 'onlineSavingsMsg'
Learn More

BD Negative Control for Caspase Inhibitors, Z-FA-FMK

A negative control inhibitor which has no inhibitory effect on apoptosis mediated by caspases and can only inhibit cysteine proteases

Brand:  BD 550411

Product Code. 15846268

This item is not returnable. View return policy

Description

Description

Members of the caspase family play key roles in inflammation and mammalian apoptosis. Z-FA-FMK is designed to be used in both in vivo and in vitro cell based assays as a negative control in assays used to measure apoptosis.

  • Contents: Lyophilized; Reconstitute in DMSO
  • Format: Purified
  • Molecular weight: 386 Daltons
  • Storage buffer: Lyophilized in dimethyl sulfoxide (DMSO)

Avoid multiple freeze-thaws of product.

Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow cytometry
-20°C
386
Purified
Lyophilized in Dimethyl Sulfoxide
1 mg
Z-FA-FMK
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.