Learn More
BD Organelle Sampler Kit
For blotting and immunoassay procedures
Brand:  BD 612740
2126.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15846279
  • 2130.00 € / Each
Explore more special offers
Alert:
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Includes: Annexin II, BiP/GRP78, b-Catenin, Caveolin 1, Connexin-43, EEA1, GM130, Integrina2/VLA-2a, Lamp-1, MAP2B, Bcl-2, Nucleoporin p62, Paxillin, ZO-1, Storage Buffer: aqueous buffered solution containing BSA, glycerol, and ≥0.09% sodium azide
Description
Description
  • Concentration: 250μg/mL
Western Blotting, Immunoprecipitation, Immunofluorescence, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Organelle Sampler Kit > 1 kit
Research use only