Learn More
BD Pharmingen™ IHC Zinc Fixative
An alternative to formalin fixation of tissue for paraffin embedded sections
Brand:  BD 550523
 View more versions of this product
Product Code. 10400275
  • 159.00 € / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.
  • Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
  • Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases
There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
1 X
1000mL
Liquid
Room temperature
Fixative
Immunohistochemical staining