Learn More
BD Pharmingen™ MitoStatus Red
In cells undergoing apoptosis, oxidative stress, necrosis, and other cellular processes, the mitochondrial membrane potential (ΔΨm) can become depolarized.
Brand:  BD 564697
97.27 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15883819
  • 97.30 € / Each
Explore more special offers
Description
Description
In cells undergoing apoptosis, pro-apoptotic Bcl-2 family proteins cause mitochondrial outer membrane permeabilization (MOMP), resulting in the release of cytochrome C and the subsequent activation of caspase-9 and the apoptotic cascade. MOMP often correlates with the loss of inner mitochondrial membrane potential (ΔΨm), which can be detected using ΔΨm-sensitive dyes. These cationic, lipophilic dyes accumulate within the mitochondria of healthy cells, but not within mitochondria that have lost ΔΨm due to induction of apoptosis or treatment with a mitochondrial uncoupler.
BD Pharmingen™ MitoStatus Red is a fluorescent dye that is readily sequestered by active mitochondria, allowing for flow cytometric or imaging analysis to assess for apoptosis or mitochondrial depolarization. Apoptotic or cells with depolarizard mitochondria will have diminished levels of fluorescence. MitoStatus Red has an excitation maximum of 622nm and an emission maximum of 648nm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 μg
Tested in Development: Human, Mouse
Upon arrival, store the dye powder desiccated and protected from light at ≤ -20°C until use. The dye powder is stable for at least 12 months if stored as indicated. After reconstitution with DMSO, store the stock solution at ≤ -20°C in small aliquots. The stock solution is stable for at least 6 months if stored as indicated.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Pharmingen™ MitoStatus Red > 100ug