missing translation for 'onlineSavingsMsg'
Learn More

BD Phosflow™ Fix Buffer I

For simultaneous fixation and permeabilization of cells prior to intracellular staining

Brand:  BD 557870


Product Code. 12830470

  • 870.00 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Contains formaldehyde and is a harmful solution.

This item is not returnable. View return policy

Description

Description

  • Contains 250mL of a 1X buffer solution
  • Optimized for use with the BD Phosflow brand of phosphorylated intracellular signaling protein-specific antibodies

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100°C
Fixation Buffer
Simultaneous fixation and permeabilization of cells prior to intracellular staining
Liquid
1 X
250mL
7.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For research use only.