Learn More
Phosflow Perm Buffer III, BD
For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood
Brand:  BD Biosciences 558050
Product Code. 15849178
  • 440.00 € / Each
Explore more special offers
Description
Description
  • Designed for optimal fluorescent staining in flow cytometry
  • Can be used with antibodies for phospho-protein analysis
  • In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Liquid
RUO
Room Temperature
125 mL
Staining Buffer
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • perm bufferr III
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : methanol