missing translation for 'onlineSavingsMsg'
Learn More

BD Port-A-Cul™ Anaerobic Collection and Transport Systems

Versatile—use for fluid, tissue, or swab specimens

33.85 € - 108.00 €

Specifications

Quantity 10 Pk.
Storage Requirements 20°C to 25°C
View More Specs

Products 4
Product Code Brand For Use With (Application) Product Type Sterility Price Quantity & Availability  
Product Code Brand For Use With (Application) Product Type Sterility Price Quantity & Availability  
17332331
missing translation for 'viewDocuments'
BD
221606
To maintain the viability of anaerobic, facultative and aerobic microorganisms during transport from the patient to the laboratory Port-A-Cul™ Tube Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17342331
missing translation for 'viewDocuments'
BD
221607
To maintain the viability of anaerobic, facultative and aerobic microorganisms during transport from the patient to the laboratory Port-A-Cul™ Tube Sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17362331
missing translation for 'viewDocuments'
BD
221609
Used for transport of fluid specimens Port-A-Cul™ Vial Sterile
91.25 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
17352331
missing translation for 'viewDocuments'
BD
221608
To maintain the viability of anaerobic, facultative and aerobic microorganisms during transport from the patient to the laboratory Port-A-Cul™ Vial Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Collect and maintain specimens at 20° to 25°C for up to 72 hours
  • Ensure aerobic, anaerobic, microaerophilic or facultative microorganism viability during transport from patient to laboratory (including Campylobacter jejuni)
  • Reduced medium formulated to protect oxygen-sensitive microbes
  • Media contain an indicator that continuously monitors the reduced media status
  • Collect specimen on a swab, as a fluid, or as a tissue specimen
  • Available as tube system (capacity 11mL), jar system (capacity 20mL), or vial system (capacity 5mL)
  • Sterile Package contains either one tube and two polyester-tipped wooden swabs, one vial or one jar
  • Screw-cap jar is 3 in. tall and has a 1 in.-wide mouth; not for use with fluids
  • Fluid Collection Kit contains one sterile-packaged vial containing media and a 5cc syringe with a 22-gauge needle and Luer-Lok™ closure

Specifications

Specifications

10 Pk.
20°C to 25°C
Videos
SDS
Documents

Documents

Certificates