Learn More
BD Propidium Iodide Staining Solution
Designed for use in two-color Annexin V flow cytometric assays.
Brand:  BD 556463
87.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15818708
  • 87.60 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detected in the orange range of the spectrum using a 562-588nm band pass filter
  • Used to assess plasma membrane (PM) integrity in Annexin V apoptosis assays
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pharmingen Propidium Iodine Staining Solution
2mL