Learn More
BD Rat C-reactive protein (CRP) ELISA Kit
For detection and quantitation of rat CRP in rat serum
Brand:  BD 557825
Product Code. 10348293
  • 1400.00 € / Each
Explore more special offers
Includes: Capture antibody coated microplate, detection antibody/enzyme conjugate (100x), standard (10x), wash buffer, TMB substrate, stop solution
Description
Description
The C-reactive protein is an acute-phase protein produced by liver during conditions of inflammation, bacterial infection, or tissue trauma
  • Reactivity: Rat
  • Useful for determination of inflammatory conditions
  • Utilizes an antibody specific for rat CRP coated on a 96-well plate
Enzyme-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRP
Rat C-Reactive protein ELISA microplate (96-well coated with Anti-CRP), Rat C-Reactive protein standard (10X), HRP Conjugated Rabbit Anti-rat CRP (100X), PBS-T Wash Buffer powder, TMB Substrate, stop solution
Rat Serum
Detection and quantitation of Rat CRP in Rat Serum, ELISA
Rat
1 Plate
2°C to 8°C
Rat C-reactive Protein ELISA kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only