missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Tumor Necrosis Factor(TNF-α)
Pure by SDS-PAGE analysis, and an absorbance assay based on the Beers-Lambert law
Brand: BD 554618
380.92 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Frozen liquid comprised of 0.22μm sterile-filtered PBS, pH 7.2, and containing 4mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity Range: Measured in a cytolysis assay using L929 indicator cells
- Specific activity: 0.05 - 2.0 X 109 Units/mg
- ED50: 5 TO 200pg/ml
- Observed Dose response: 100 fold
Bioassay. enzyme-linked immunosorbent assay standard

Specifications
200μg/mL | |
Frozen aqueous Buffered solution containing BSA | |
10μg | |
Human |
Bioassay, ELISA Standard, ELISA | |
Human TNF-a Recombinant Protein | |
-80°C | |
Recombinant |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Recombinant Human Tumor Necrosis Factor(TNF-α) > Recombinant Human Tumor Necrosis Factor(TNF-alpha); 200μg/mL; 10μg
Research Use Only
Spot an opportunity for improvement?Share a Content Correction