missing translation for 'onlineSavingsMsg'
Learn More

BD Retrievagen Antigen Retrieval Systems

Product Code. 15806308
Click to view available options
Quantity:
125 mL
250 mL
Reagent Type:
Retrievagen A
Retrievagen B
Unit Size:
1 L
Each
This item is not returnable. View return policy

Product Code. 15806308

Brand: BD 550527

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining

10% neutral buffered formalin fixes the tissue by cross-linking proteins that may cause a conformational change or “masking” of the antigenic epitope leading to reduced or absent antibody reactivity. The BD Retrievagen Antigen Retrieval Systems are designed to unmask the epitope and cause the enhancement of antibody staining.

  • Retrievagen A (pH 6.0) is provided as two separate solutions that need to be mixed
  • Retrievagen B (pH 9.5) is provided as a ready-to-use solution

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Tris based Buffer with chemical agents for epitope retrieval
Product Type Retrieval Reagent
Quantity 250 mL
For Use With (Application) Immunohistochemical staining
pH 9.5
Regulatory Status RUO
Reagent Type Retrievagen B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.