Learn More
BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
Brand:  BD 367323
 View more versions of this product
Product Code. 12937746
  • 374.00 € / Pack of 200
Explore more special offers
Description
Description
  • Ease of use reduces training requirements and improves activation compliance
  • In-vein activation reduces the risk exposure to a contaminated needle
  • Easy activation without additional patient discomfort
  • Ideal for use in high-risk environments No luer adapter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Push Button Blood Collection Set
25 G
3/4 in.
12 in.