Learn More
BD Green Nucleic Acid Stain
Useful for discrimination of viable from nonviable cells, DNA content analysis in fixed cells, or nuclear counterstaining for fixed cells in immunofluorescence or flow cytometry applications. BD Via-Probe™ Green Nucleic Acid Stain brightly stains nonviable or fixed cells with permeable membranes.
Brand:  BD 565799
 View more versions of this product
Product Code. 15884529
  • 176.00 € / Each
Explore more special offers
Description
Description
  • Green Nucleic Acid Stain has an excitation maximum of 503nm, and is best excited by the blue laser
  • It has an emission maximum of 526nm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-20°C
  • Immunofluorescence
  • Immunocytochemistry
  • Flow cytometry
  • Intracellular staining (flow cytometry)
Green Nucleic Acid Stain
0.1 mL
Green