missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15417-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the short-chain dehydrogenase/reductase gene family. The encoded protein forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific requirement for phosphatidylcholine for optimal enzymatic activity. The encoded protein catalyzes the interconversion of acetoacetate and -3-hydroxybutyrate, the two major ketone bodies produced during fatty acid catabolism. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
BDH1 | |
Polyclonal | |
Unconjugated | |
BDH1 | |
BDH, BDH1, SDR9C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117099, 622, 71911 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29147, Q02338, Q80XN0 | |
BDH1 | |
BDH1 Fusion Protein Ag7705 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |