Learn More
Pinnacle Brush™ Beaker Brushes
Constructed on galvanised steel wire handle. Pinnacle Brush™ Beaker Brushes have nylon bristles for gentle cleaning.
Brand:  Pinnacle Brush™ T309
 View more versions of this product
Product Code. 12811934
  • 220.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Galvanized steel handle
  • Nylon bristles
Specifications
Specifications
Bottle Brush
Metal
27 cm
17.5 cm
Brush Circumference: 14 cm
Fiber
4.5 cm
9.5 cm
10 Pk.