missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Beatus Bottle Top Dispenser
Robust and reliable dispensing; Fisherbrand™ Beatus Bottle Top Dispenser has re-circulation valve for re-directing the liquid into the bottle for bubble free dispensing without any loss of reagent.
Brand: Fisherbrand™ BEAT100
Description
- With re-circulation valve
- Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
- Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
- Specially designed adaptor allows 360° rotation providing full flexibility
- Springless, PTFE valve manifold ensures high chemical compatibility and jamming free valve function
- PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
- Adaptors to fit most of the laboratory reagent bottles
- Calibration tool provided for convenient and quick in-lab user re-calibration
Specifications:
- Piston: PTFE and ETP
- Valve housing: PFA
- Re-Circulation valve housing: PFA
- Valve assembly: Borosilicate glass and seat
- Discharge assembly: PTFE
- Delivery and Inlet tube: FEP
- Compatibility: With all reagents except HF
Specifications
0.6% (+/-0.600) | |
Autoclavable (121°C, 15psi for 10-15 min.) | |
Borosilicate Glass | |
2.0 mL |
Bottletop Dispenser | |
ISO 8655 standards | |
10 to 100 mL | |
BEAT-100 |
Videos
-
Talking Points - Fisherbrand Bottle Top Dispensers
Instruction manual accumet AE6
File size: 4.9 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Beatus Bottle Top Dispenser > Increment: 2.0mL; Volume: 10 - 100mL
Spot an opportunity for improvement?Share a Content Correction